Please login to your online account to display your discounted pricing

N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine, 96%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  34430-0010

 View more versions of this product

Catalog No. AC344300010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Quantity 1g
Color Clear colorless to light yellow
Chemical Name or Material N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
Infrared Spectrum Authentic
Assay Percent Range 96%
Linear Formula C6H5CH2N(CH2OCH3)CH2Si(CH3)3
MDL Number MFCD00674005
CAS 93102-05-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Molecular Formula C13H23NOSi
Elemental Analysis 95% min.
CAS Min % 95.0
Health Hazard 1 Warning
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4910 to 1.4940 (20°C, 589nm)
Formula Weight 237.42
Density 0.9280g/mL

Specifications

Chemical Name or Material N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
CAS 93102-05-7
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C13H23NOSi
Linear Formula C6H5CH2N(CH2OCH3)CH2Si(CH3)3
Formula Weight 237.42
MDL Number MFCD00674005
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents