missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methoxy-N-methylbenzamide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436560250

 View more versions of this product

Catalog No. AC436560250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Methoxy-N-methylbenzamide
Authentic
85°C (1.0mmHg)
25g
97.5
97.5% min. (GC)
acmc-1b5bp
CN(C(=O)C1=CC=CC=C1)OC
165.19
CHEBI:59742
Liquid
113°C
Glass bottle
Colorless to Yellow
6919-61-5
100.0
C9H11NO2
UKERDACREYXSIV-UHFFFAOYSA-N
N-methoxy-N-methylbenzamide
569575
165.19
98%

Chemical Identifiers

6919-61-5
165.19
acmc-1b5bp
CHEBI:59742
CN(C(=O)C1=CC=CC=C1)OC
C9H11NO2
UKERDACREYXSIV-UHFFFAOYSA-N
569575
N-methoxy-N-methylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Documents