missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methoxy-N-methylbenzamide, 98%

Manufacturer:  Alfa Aesar™ H5425306

 View more versions of this product

Catalog No. AAH5425306



Specifications

Specifications

N-Methoxy-N-methylbenzamide
70°C
6919-61-5
MFCD00075320
UKERDACREYXSIV-UHFFFAOYSA-N
N-methoxy-N-methylbenzamide
569575
165.19
113°C (235°F)
5g
C9H11NO2
acmc-1b5bp
CN(C(=O)C1=CC=CC=C1)OC
165.192
CHEBI:59742
98%

Chemical Identifiers

6919-61-5
165.192
UKERDACREYXSIV-UHFFFAOYSA-N
569575
N-methoxy-N-methylbenzamide
C9H11NO2
MFCD00075320
acmc-1b5bp
CHEBI:59742
CN(C(=O)C1=CC=CC=C1)OC
Documents