missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methoxy-N-methylacetamide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC369310050

 View more versions of this product

Catalog No. AC369310050



Description

Description

Specifications

Specifications

N-Methoxy-N-methylacetamide
49°C
Glass bottle
0.97
Colorless
98%
97.5
97.5% min. (GC)
CH3CON(OCH3)CH3
acetamide, n-methoxy-n-methyl, acmc-209pdo, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamde, n-methoxy-n-methylacetamid, n-methoxyl-n-methylacetamide, n-methyl-n-methoxyacetamide, pubchem12775
OYVXVLSZQHSNDK-UHFFFAOYSA-N
N-methoxy-N-methylacetamide
537505
Liquid
0.9700g/mL
Authentic
1.4240 to 1.426
152°C
5g
78191-00-1
100.0
C4H9NO2
MFCD00060098
Solubility in water: soluble
CON(C)C(C)=O
103.12
103.12
98%

Chemical Identifiers

78191-00-1
103.12
OYVXVLSZQHSNDK-UHFFFAOYSA-N
537505
CON(C)C(C)=O
C4H9NO2
MFCD00060098
acetamide, n-methoxy-n-methyl, acmc-209pdo, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamde, n-methoxy-n-methylacetamid, n-methoxyl-n-methylacetamide, n-methyl-n-methoxyacetamide, pubchem12775
N-methoxy-N-methylacetamide
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents