Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Methoxy-N-methylacetamide, 98%

Chemical Identifiers

Chemical Name or Material N-Methoxy-N-methylacetamide
CAS 78191-00-1
Formula Weight 103.12
MDL Number MFCD00060098
UN Number UN1993
Beilstein 3600878
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6522606

  SDS
alfa aesar™
H6522606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6522614

  SDS
alfa aesar™
H6522614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6522622

  SDS
alfa aesar™
H6522622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 103.12
CAS 78191-00-1
Chemical Name or Material N-Methoxy-N-methylacetamide
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
MDL Number MFCD00060098
Assay 98%
Physical Form Liquid
UN Number UN1993
Density 0.970g/mL
Beilstein 3600878
Health Hazard 1 Warning
Boiling Point 152°C
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Refractive Index 1.4260
Color Colorless
Quantity 5g
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.