missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methoxy-N-methyl-4-(trifluoromethyl)benzamide, TCI America™

$204.72 - $710.44

Chemical Identifiers

CAS 116332-61-7
Molecular Formula C10H10F3NO2
Molecular Weight (g/mol) 233.19
MDL Number MFCD08689795
InChI Key YFDOMHRDNWBTMU-UHFFFAOYSA-N
PubChem CID 14069348
IUPAC Name N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
SMILES CN(C(=O)C1=CC=C(C=C1)C(F)(F)F)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27625G View Documents TCI America
M27625G
5g Each for $710.44
 
M27621G View Documents TCI America
M27621G
1g Each for $204.72
 
Specifications

Chemical Identifiers

116332-61-7
233.19
YFDOMHRDNWBTMU-UHFFFAOYSA-N
N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
C10H10F3NO2
MFCD08689795
14069348
CN(C(=O)C1=CC=C(C=C1)C(F)(F)F)OC

Specifications

Yellow
116332-61-7
MFCD08689795
CN(C(=O)C1=CC=C(C=C1)C(F)(F)F)OC
233.19
233.19
≥98.0% (GC)
5g
C10H10F3NO2
YFDOMHRDNWBTMU-UHFFFAOYSA-N
N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
14069348
Clear Liquid at 20°C
N-Methoxy-N-methyl-4-(trifluoromethyl)benzamide
Safety and Handling

Safety and Handling

TSCA : No