Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Linoleoylglycine, 98%

Chemical Identifiers

Chemical Name or Material N-Linoleoylglycine
CAS 2764-03-6
Formula Weight 337.5
MDL Number MFCD11977729
Synonym LA-Gly
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67453LB0

 
alfa aesar™
J67453LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67453MA

 
alfa aesar™
J67453MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 337.5
Chemical Name or Material N-Linoleoylglycine
Physical Form Solid
Assay 98%
MDL Number MFCD11977729
CAS 2764-03-6
Synonym LA-Gly
Quantity 5mg
Storage Note 1 -30° to -10°C
RTECS Number MC0596320
Safety and Handling

-30° to -10°C