missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Linoleoylglycine, 98%

Manufacturer:  Alfa Aesar™ J67453MA

 View more versions of this product

Catalog No. AAJ67453MA



Specifications

Specifications

N-Linoleoylglycine
C20H35NO3
glycine linoleamide, glycine, n-1-oxo-9,12-octadecadienyl-, z,z, glycine, n-linoleoyl, glycine-linoleamide, linoleamide de glycine french, linoleoyl glycine, linoleoylglycine, linolyl glycine, n-linoleoyl-glycine, n-linoleoylglycine
CCCCCC=CCC=CCCCCCCCC(=O)NCC(=O)O
337.50
337.5
98%
2764-03-6
MFCD11977729
YCRHZEHWEYAHCO-HZJYTTRNSA-N
2-[[(9Z,12Z)-octadeca-9,12-dienoyl]amino]acetic acid
6433346
Solid
10mg

Chemical Identifiers

2764-03-6
337.50
YCRHZEHWEYAHCO-HZJYTTRNSA-N
6433346
CCCCCC=CCC=CCCCCCCCC(=O)NCC(=O)O
C20H35NO3
MFCD11977729
glycine linoleamide, glycine, n-1-oxo-9,12-octadecadienyl-, z,z, glycine, n-linoleoyl, glycine-linoleamide, linoleamide de glycine french, linoleoyl glycine, linoleoylglycine, linolyl glycine, n-linoleoyl-glycine, n-linoleoylglycine
2-[[(9Z,12Z)-octadeca-9,12-dienoyl]amino]acetic acid
Safety and Handling

Safety and Handling

RTECSNumber : MC0596320

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents