missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Lauroylsarcosine, TCI America™

$27.50 - $174.17

Chemical Identifiers

CAS 97-78-9
Molecular Formula C15H29NO3
Molecular Weight (g/mol) 271.401
MDL Number MFCD00021749
InChI Key BACYUWVYYTXETD-UHFFFAOYSA-N
Synonym n-lauroylsarcosine, lauroylsarcosine, n-dodecanoylsarcosine, lauroyl sarcosine, hamposyl l, maprosyl l, sarcosyl l, n-laurylsarcosine, n-dodecanoyl-n-methylglycine, glycine, n-methyl-n-1-oxododecyl
PubChem CID 7348
IUPAC Name 2-[dodecanoyl(methyl)amino]acetic acid
SMILES CCCCCCCCCCCC(=O)N(C)CC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L015125G View Documents TCI America
L015125G
25g Each for $27.50
 
L0151500G View Documents TCI America
L0151500G
500g Each for $174.17
 
L0151100G View Documents TCI America
L0151100G
100g Each for $65.70
 
Specifications

Chemical Identifiers

97-78-9
271.401
BACYUWVYYTXETD-UHFFFAOYSA-N
7348
CCCCCCCCCCCC(=O)N(C)CC(=O)O
C15H29NO3
MFCD00021749
n-lauroylsarcosine, lauroylsarcosine, n-dodecanoylsarcosine, lauroyl sarcosine, hamposyl l, maprosyl l, sarcosyl l, n-laurylsarcosine, n-dodecanoyl-n-methylglycine, glycine, n-methyl-n-1-oxododecyl
2-[dodecanoyl(methyl)amino]acetic acid

Specifications

46°C
97-78-9
MFCD00021749
BACYUWVYYTXETD-UHFFFAOYSA-N
2-[dodecanoyl(methyl)amino]acetic acid
7348
≥90.0% (GC)
25g
C15H29NO3
n-lauroylsarcosine, lauroylsarcosine, n-dodecanoylsarcosine, lauroyl sarcosine, hamposyl l, maprosyl l, sarcosyl l, n-laurylsarcosine, n-dodecanoyl-n-methylglycine, glycine, n-methyl-n-1-oxododecyl
CCCCCCCCCCCC(=O)N(C)CC(=O)O
271.401
271.40
N-Lauroylsarcosine
Safety and Handling

Safety and Handling

TSCA : Yes