Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Isopropylthiourea, 97%

$65.69 - $216.97

Chemical Identifiers

Chemical Name or Material N-Isopropylthiourea
CAS 1719-76-2
Molecular Formula C4H10N2S
Formula Weight 118.2
MDL Number MFCD00041375
Beilstein 1743052
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1260503

  SDS
alfa aesar™
L1260503
1g Each for $65.69

AAL1260506

  SDS
alfa aesar™
L1260506
5g Each for $216.97
Description & Specifications

Specifications

Formula Weight 118.2
CAS 1719-76-2
Chemical Name or Material N-Isopropylthiourea
MDL Number MFCD00041375
Beilstein 1743052
Assay 97%
Melting Point 166° to 170°C
Health Hazard 1 Warning
RTECS Number YT6125000
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Molecular Formula C4H10N2S
Odor Odorless
Quantity 1g
Health Hazard 3 GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
N-Isopropylthiourea is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.