missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Isopropylthiourea, 97%

Manufacturer:  Alfa Aesar™ L1260503

 View more versions of this product

Catalog No. AAL1260503



Description

Description

Applications
N-Isopropylthiourea is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

N-Isopropylthiourea
166°C to 170°C
1719-76-2
MFCD00041375
n-isopropylthiourea, isopropylthiourea, isopropyl thiourea, urea, 1-isopropyl-2-thio, usaf d-6, 1-isopropyl-2-thiourea, isopropyl-thiourea, 1-isopropylthiourea, propan-2-yl thiourea, 1-iso-propyl-2-thiourea
CC(C)NC(=S)N
118.198
118.2
Odorless
1g
C4H10N2S
1743052
POXAIQSXNOEQGM-UHFFFAOYSA-N
propan-2-ylthiourea
1711921
97%

Chemical Identifiers

1719-76-2
118.198
POXAIQSXNOEQGM-UHFFFAOYSA-N
1711921
CC(C)NC(=S)N
C4H10N2S
MFCD00041375
n-isopropylthiourea, isopropylthiourea, isopropyl thiourea, urea, 1-isopropyl-2-thio, usaf d-6, 1-isopropyl-2-thiourea, isopropyl-thiourea, 1-isopropylthiourea, propan-2-yl thiourea, 1-iso-propyl-2-thiourea
propan-2-ylthiourea
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YT6125000

Documents