Learn More
N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6589214
Description
N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.
Solubility
Soluble in chloroform, DMSO, methanol.
Notes
Air Sensitive. Store away from air. Store under dry inert gas. Do not store together with acids. Incompatible with acids , air and oxidizing agents.
Specifications
N-Isopropylmethylamine | |
Colorless | |
50°C to 53°C | |
C4H11N | |
(CH3)2CHNHCH3 | |
25 g | |
1730877 | |
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine | |
XHFGWHUWQXTGAT-UHFFFAOYSA-N | |
N-methylpropan-2-amine | |
78485 | |
98% |
4747-21-1 | |
0.702 g/mL | |
-32°C (-26°F) | |
1.348 | |
MFCD00042859 | |
UN2734 | |
Air Sensitive | |
Soluble in chloroform,DMSO,methanol. | |
CC(C)NC | |
73.139 | |
73.14 | |
Liquid |
Chemical Identifiers
4747-21-1 | |
73.139 | |
XHFGWHUWQXTGAT-UHFFFAOYSA-N | |
78485 | |
CC(C)NC |
C4H11N | |
MFCD00042859 | |
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine | |
N-methylpropan-2-amine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H301+H331-H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: I; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber : 225-266-7
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only