Learn More
N-Isopropylhydroxylamine hydrochloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 276260050
Specifications
N-Isopropylhydroxylamine hydrochloride | |
Glass bottle | |
68°C to 72°C | |
97% | |
96.0 | |
96% min. (Argentometry) | |
(CH3)2CHNHOH·HCl | |
04, 538 | |
BYXUIKZQGOPKFR-UHFFFAOYSA-N | |
N-propan-2-ylhydroxylamine;hydrochloride | |
170873 | |
Adhering Crystalline Powder |
Authentic | |
White to Beige | |
5g | |
50632-53-6 | |
100.0 | |
C3H9NO·HCl | |
MFCD00012599 | |
2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride, n-isopropylhydroxylamine hcl, n-isopropylhydroxylamine hydrochloride | |
CC(C)NO.Cl | |
111.57 | |
111.57 | |
97% |
Chemical Identifiers
50632-53-6 | |
111.57 | |
BYXUIKZQGOPKFR-UHFFFAOYSA-N | |
170873 | |
CC(C)NO.Cl |
C3H9NO·HCl | |
MFCD00012599 | |
2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride, n-isopropylhydroxylamine hcl, n-isopropylhydroxylamine hydrochloride | |
N-propan-2-ylhydroxylamine;hydrochloride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning