missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Isopropylethylenediamine, TCI America™

$206.25 - $400.28

Chemical Identifiers

CAS 19522-67-9
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00008164
InChI Key KDRUIMNNZBMLJR-UHFFFAOYSA-N
Synonym n-isopropylethylenediamine, 2-isopropylaminoethylamine, isopropylaminoethylamine, 1,2-ethanediamine, n-1-methylethyl, ethylenediamine, n-isopropyl, n-isopropylethane-1,2-diamine, 2-aminoethyl propan-2-yl amine, 1,2-ethanediamine, n1-1-methylethyl, 2-aminoethyl isopropyl amine, n1-isopropylethane-1,2-diamine
PubChem CID 88098
ChEBI CHEBI:84288
IUPAC Name N'-propan-2-ylethane-1,2-diamine
SMILES CC(C)NCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I038110ML View Documents TCI America
I038110ML
10mL Each for $206.25
 
I038125ML View Documents TCI America
I038125ML
25mL Each for $400.28
 
Specifications

Chemical Identifiers

19522-67-9
102.181
KDRUIMNNZBMLJR-UHFFFAOYSA-N
88098
N'-propan-2-ylethane-1,2-diamine
C5H14N2
MFCD00008164
n-isopropylethylenediamine, 2-isopropylaminoethylamine, isopropylaminoethylamine, 1,2-ethanediamine, n-1-methylethyl, ethylenediamine, n-isopropyl, n-isopropylethane-1,2-diamine, 2-aminoethyl propan-2-yl amine, 1,2-ethanediamine, n1-1-methylethyl, 2-aminoethyl isopropyl amine, n1-isopropylethane-1,2-diamine
CHEBI:84288
CC(C)NCCN

Specifications

138°C
10mL
C5H14N2
2733
KDRUIMNNZBMLJR-UHFFFAOYSA-N
N'-propan-2-ylethane-1,2-diamine
88098
102.18
≥98.0% (GC,T)
Yellow
19522-67-9
MFCD00008164
n-isopropylethylenediamine, 2-isopropylaminoethylamine, isopropylaminoethylamine, 1,2-ethanediamine, n-1-methylethyl, ethylenediamine, n-isopropyl, n-isopropylethane-1,2-diamine, 2-aminoethyl propan-2-yl amine, 1,2-ethanediamine, n1-1-methylethyl, 2-aminoethyl isopropyl amine, n1-isopropylethane-1,2-diamine
CC(C)NCCN
102.181
CHEBI:84288
Clear Liquid at 20°C
N-Isopropylethylenediamine
Safety and Handling

Safety and Handling

TSCA : Yes