Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Isopropylcarbodiimide, 0.5-0.8 mmol/g on Merrifield resin

Chemical Identifiers

Chemical Name or Material N-Isopropylcarbodiimide
Name Note 0.5 to 0.8mmol/g on Merrifield resin
MDL Number MFCD03426036
Synonym N-Isopropylcarbodiimide, N'-methyl polystyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1946203

 
alfa aesar™
L1946203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material N-Isopropylcarbodiimide
Quantity 1g
Name Note 0.5 to 0.8mmol/g on Merrifield resin
MDL Number MFCD03426036
Synonym N-Isopropylcarbodiimide, N'-methyl polystyrene