Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Isopropyl-N'-phenyl-p-phenylenediamine, 95%

Chemical Identifiers

Chemical Name or Material N-Isopropyl-N'-phenyl-p-phenylenediamine
CAS 101-72-4
Molecular Formula C15H18N2
Formula Weight 226.32
MDL Number MFCD00026348
UN Number UN3077
Synonym 4-(Isopropylamino)diphenylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2537830

  SDS
alfa aesar™
B2537830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB25378A1

  SDS
alfa aesar™
B25378A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.32
Health Hazard 3 GHS P Statement
P280g-P262-P273-P501a
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00026348
Health Hazard 1 Warning
Flash Point 172°C (342°F)
Assay 95%
TSCA TSCA
Synonym 4-(Isopropylamino)diphenylamine
CAS 101-72-4
Boiling Point 161°C (1mmHg)
UN Number UN3077
RTECS Number ST2650000
Quantity 250g
Molecular Formula C15H18N2
Melting Point 74° to 80°C
Health Hazard 2 GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Chemical Name or Material N-Isopropyl-N'-phenyl-p-phenylenediamine
Safety and Handling

Warning

GHS P Statement
P280g-P262-P273-P501a
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.