Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Isopropyl-2-chloroacetamide, 97%

$62.68 - $197.39

Chemical Identifiers

Chemical Name or Material N-Isopropyl-2-chloroacetamide
CAS 2895-21-8
Molecular Formula ClCH2CONHCH(CH3)2
Formula Weight 135.59
MDL Number MFCD00068034
Beilstein 1745034
Synonym N-(Chloroacetyl)isopropylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2486303

  SDS
alfa aesar™
B2486303
1g Each for $62.68

AAB2486306

  SDS
alfa aesar™
B2486306
5g Each for $197.39
Description & Specifications

Specifications

Formula Weight 135.59
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 58° to 60°C
MDL Number MFCD00068034
Molecular Formula ClCH2CONHCH(CH3)2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2895-21-8
Quantity 1g
Chemical Name or Material N-Isopropyl-2-chloroacetamide
Health Hazard 1 Warning
Synonym N-(Chloroacetyl)isopropylamine
Beilstein 1745034
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.