Please login to your online account to display your discounted pricing

N-Isopropyl-1-piperazineacetamide, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  205610050

 View more versions of this product

Catalog No. AC205610050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystals and Chunks
Formula Weight 185.27
Color Slightly yellow
Quantity 5g
EINECS Number 254-676-9
Melting Point 82° to 88°C
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material N-Isopropyl-1-piperazineacetamide
Name Note 95%
Assay Percent Range 95%
Molecular Formula C9H19N3O
MDL Number MFCD00005969
CAS 39890-42-1

Specifications

Chemical Name or Material N-Isopropyl-1-piperazineacetamide
Name Note 95%
CAS 39890-42-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H19N3O
Formula Weight 185.27
MDL Number MFCD00005969