Please login to your online account to display your discounted pricing

N-Isopropyl-1-piperazineacetamide, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Isopropyl-1-piperazineacetamide
Name Note 95%
CAS 39890-42-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C9H19N3O
Formula Weight 185.27
MDL Number MFCD00005969
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC205610050

  SDS
fisher scientific acros
205610050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Color Slightly yellow
Physical Form Crystals and Chunks
Formula Weight 185.27
Quantity 5g
EINECS Number 254-676-9
Melting Point 82° to 88°C
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 95%
MDL Number MFCD00005969
Name Note 95%
Chemical Name or Material N-Isopropyl-1-piperazineacetamide
Molecular Formula C9H19N3O
CAS 39890-42-1