Please login to your online account to display your discounted pricing

N-Iodosuccinimide, 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC299575000

 View more versions of this product

Catalog No. AC299575000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5% (Iodometry)
Formula Weight 224.99
Quantity 500g
Color White-yellow to brown
Melting Point 197° to 204°C
Chemical Name or Material N-Iodosuccinimide
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C4H4INO2
CAS 516-12-1

Specifications

Chemical Name or Material N-Iodosuccinimide
Assay ≥97.5% (Iodometry)
Assay Percent Range 98%
Molecular Formula C4H4INO2
Formula Weight 224.99