missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Iodosuccinimide, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1432022

 View more versions of this product

Catalog No. AAA1432022



Description

Description

Applications
N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

Solubility
Soluble in dioxane, tetrahydrfuran and acetonitrile. Insoluble in ether and carbon tetrachloride.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

N-Iodosuccinimide
100g
97%
n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
C1CC(=O)N(C1=O)I
224.985
CHEBI:53204
2.245
516-12-1
C4H4INO2
LQZMLBORDGWNPD-UHFFFAOYSA-N
1-iodopyrrolidine-2,5-dione
120273
224.99

Chemical Identifiers

516-12-1
224.985
n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
CHEBI:53204
C1CC(=O)N(C1=O)I
C4H4INO2
LQZMLBORDGWNPD-UHFFFAOYSA-N
120273
1-iodopyrrolidine-2,5-dione
Documents