Please login to your online account to display your discounted pricing

N-Iodosaccharin 95%, ACROS Organics™

$234.20 - $234.20

Chemical Identifiers

Chemical Name or Material N-Iodosaccharin
CAS 86340-94-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H4INO3S
Formula Weight 309.08
MDL Number MFCD09037939
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388340010

  SDS
thermo fisher scientific
AC388340010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC388340050

  SDS
thermo fisher scientific
AC388340050
Glass bottle 5g Each for $234.20
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 309.08
Physical Form Crystalline Powder
Quantity 1g
Color Beige
Merck Index 15, 5086
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
Chemical Name or Material N-Iodosaccharin
MDL Number MFCD09037939
Molecular Formula C7H4INO3S
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS 86340-94-5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

C-X bond formation reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.