missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Hydroxyurethane, TCI America™

$53.47 - $142.09

Chemical Identifiers

CAS 589-41-3
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00002108
InChI Key VGEWEGHHYWGXGG-UHFFFAOYSA-N
Synonym n-hydroxyurethane, ethyl hydroxycarbamate, n-hydroxyurethan, hydroxyurethane, hydroxycarbamic acid ethyl ester, n-carbethoxyhydroxylamine, n-hydroxy ethyl carbamate, hydroxyurethan, carbamic acid, hydroxy-, ethyl ester, ethyl-n-hydroxycarbamate
PubChem CID 11510
IUPAC Name ethyl N-hydroxycarbamate
SMILES CCOC(=O)NO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H06185G View Documents TCI America
H06185G
5g Each for $142.09
 
H06181G View Documents TCI America
H06181G
1g Each for $53.47
 
Specifications

Chemical Identifiers

589-41-3
105.093
VGEWEGHHYWGXGG-UHFFFAOYSA-N
11510
CCOC(=O)NO
C3H7NO3
MFCD00002108
n-hydroxyurethane, ethyl hydroxycarbamate, n-hydroxyurethan, hydroxyurethane, hydroxycarbamic acid ethyl ester, n-carbethoxyhydroxylamine, n-hydroxy ethyl carbamate, hydroxyurethan, carbamic acid, hydroxy-, ethyl ester, ethyl-n-hydroxycarbamate
ethyl N-hydroxycarbamate

Specifications

113°C
5g
C3H7NO3
n-hydroxyurethane, ethyl hydroxycarbamate, n-hydroxyurethan, hydroxyurethane, hydroxycarbamic acid ethyl ester, n-carbethoxyhydroxylamine, n-hydroxy ethyl carbamate, hydroxyurethan, carbamic acid, hydroxy-, ethyl ester, ethyl-n-hydroxycarbamate
CCOC(=O)NO
105.093
105.09
≥85.0% (GC,N)
Yellow
589-41-3
MFCD00002108
VGEWEGHHYWGXGG-UHFFFAOYSA-N
ethyl N-hydroxycarbamate
11510
Clear Liquid at 20°C
N-Hydroxyurethane
Safety and Handling

Safety and Handling

RTECSNumber : FB1750000

TSCA : No