Learn More
Thermo Scientific Chemicals N-Hydroxysulfosuccinimide sodium salt, 95%, Thermo Scientific Chemicals
Coupling reagent
Supplier: Thermo Scientific Chemicals 438650010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentSpecifications
N-Hydroxysulfosuccinimide sodium salt | |
96.0 | |
White | |
21.46 to 23.02% C, 6.19 to 6.71% N | |
Authentic | |
Glass bottle | |
n-hydroxysulfosuccinimide sodium salt, sulfo-nhs, sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate, n-hydroxysulfosuccinimidesodiumsalt, 3-pyrrolidinesulfonic acid, 1-hydroxy-2,5-dioxo-, monosodium salt, sodium 1-hydroxy-2,5-dioxo-pyrrolidine-3-sulfonate, n-hydroxusulfosuccinimide sodium, n-hydroxy-2-sodiosulfo succinimide, n-hydroxysulfosuccinimide,sodium salt, n-hydroxusulfosuccinimide sodium salt | |
C1C(C(=O)N(C1=O)O)S(=O)(=O)[O-].[Na+] | |
217.13 | |
217.13 | |
95% |
106627-54-7 | |
100.0 | |
250.0°C | |
95% | |
C4H4NNaO6S | |
1g | |
RPENMORRBUTCPR-UHFFFAOYSA-M | |
sodium;1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate | |
3520574 | |
Powder |
Chemical Identifiers
106627-54-7 | |
217.13 | |
n-hydroxysulfosuccinimide sodium salt, sulfo-nhs, sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate, n-hydroxysulfosuccinimidesodiumsalt, 3-pyrrolidinesulfonic acid, 1-hydroxy-2,5-dioxo-, monosodium salt, sodium 1-hydroxy-2,5-dioxo-pyrrolidine-3-sulfonate, n-hydroxusulfosuccinimide sodium, n-hydroxy-2-sodiosulfo succinimide, n-hydroxysulfosuccinimide,sodium salt, n-hydroxusulfosuccinimide sodium salt | |
sodium;1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate |
C4H4NNaO6S | |
RPENMORRBUTCPR-UHFFFAOYSA-M | |
3520574 | |
C1C(C(=O)N(C1=O)O)S(=O)(=O)[O-].[Na+] |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
If skin ir
GHS Signal Word: Warning
RUO â Research Use Only