Please login to your online account to display your discounted pricing

N'-Hydroxypyridine-4-carboximidamide, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N'-Hydroxypyridine-4-carboximidamide
CAS 1594-57-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H7N3O
Formula Weight 137.14
MDL Number MFCD00125873
Synonym 4-Pyridylamidoxime
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432480050

  SDS
thermo fisher scientific
AC432480050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC432480010

  SDS
thermo fisher scientific
AC432480010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Solid
Assay ≥96.0%
Formula Weight 137.14
Quantity 5g
Color White to off-white
Melting Point 200° to 207°C
CAS Min % 96.0
CAS Max % 100.0
Synonym 4-Pyridylamidoxime
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material N'-Hydroxypyridine-4-carboximidamide
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C6H7N3O
MDL Number MFCD00125873
CAS 1594-57-6
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.