missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxyphthalimide 99.0+%, TCI America™

Supplier:  TCI America H0395500G

 View more versions of this product

Catalog No. H0395500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Hydroxyphthalimide
231°C
C8H5NO3
500 g
CFMZSMGAMPBRBE-UHFFFAOYSA-N
2-hydroxy-2,3-dihydro-1H-isoindole-1,3-dione
10665
≥99.0% (T)
524-38-9
White-Yellow
MFCD00005891
n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
ON1C(=O)C2=CC=CC=C2C1=O
163.13
163.13
Crystalline Powder

Chemical Identifiers

524-38-9
163.13
CFMZSMGAMPBRBE-UHFFFAOYSA-N
10665
ON1C(=O)C2=CC=CC=C2C1=O
C8H5NO3
MFCD00005891
n-hydroxyphthalimide, 2-hydroxyisoindoline-1,3-dione, 2-hydroxy-1h-isoindole-1,3 2h-dione, nhpi, 2-hydroxyphthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxy, phthalimide, n-hydroxy, phthaloxime, unii-bxi99m81x0, 2-hydroxyisoindolin-1,3-dione
2-hydroxy-2,3-dihydro-1H-isoindole-1,3-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6724

RTECSNumber : TI5200000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions