Please login to your online account to display your discounted pricing

N-Hydroxyphthalimide, 98%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  329871000

 View more versions of this product

Catalog No. AC329871000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 163.13
Quantity 100g
Color Yellow
EINECS Number 208-358-1
TSCA TSCA
Assay 97.5 to 102.5% (on anhydrous substance)
RTECS Number TI5200000
CAS Max % 100.0
Packaging Plastic bottle
Water 2% max.
CAS Min % 97.5
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material N-Hydroxyphthalimide
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H5NO3
MDL Number MFCD00005891
CAS 524-38-9
Synonym Phthaloxime
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material N-Hydroxyphthalimide
Name Note 98%
CAS 524-38-9
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 102.5% (on anhydrous substance)
Assay Percent Range 98%
Molecular Formula C8H5NO3
Formula Weight 163.13
MDL Number MFCD00005891
Synonym Phthaloxime
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents