Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Hydroxymethyl)trifluoroacetamide, 98+%


Quantity 25g

Manufacturer: alfa aesar™  L0012814

 View more versions of this product

Catalog No. AAL0012814

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 143.06
Chemical Name or Material N-(Hydroxymethyl)trifluoroacetamide
Assay ≥98%
Molecular Formula C3H4F3NO2
MDL Number MFCD00014416
CAS 50667-69-1
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Linear Formula CF3CONHCH2OH
Beilstein 1859914
Synonym Trifluoroacetamidomethanol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Melting Point 99° to 103°C
Quantity 25g

Specifications

Chemical Name or Material N-(Hydroxymethyl)trifluoroacetamide
CAS 50667-69-1
Molecular Formula C3H4F3NO2
Linear Formula CF3CONHCH2OH
Formula Weight 143.06
MDL Number MFCD00014416
Beilstein 1859914
Synonym Trifluoroacetamidomethanol
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C

SDS & Safety Documents