Please login to your online account to display your discounted pricing

N-(Hydroxymethyl)phthalimide 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-(Hydroxymethyl)phthalimide
Name Note 97%
CAS 118-29-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H7NO3
Formula Weight 177.16
MDL Number MFCD00005899
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121490250

  SDS
fisher scientific acros
121490250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC121491000

  SDS
fisher scientific acros
121491000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Fine Crystalline Powder
Formula Weight 177.16
Quantity 25g
Color White
Beilstein 21, 475
Fieser 01,484; 08,253
EINECS Number 204-241-4
TSCA TSCA
RTECS Number TI5270000
Melting Point 140° to 145°C
CAS Min % 96.0
CAS Max % 100.0
Loss on Drying 2% max.
Packaging Glass bottle
Sulfated Ash 0.1% max.
CAS 118-29-6
Infrared Spectrum Authentic
Chemical Name or Material N-(Hydroxymethyl)phthalimide
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H7NO3
MDL Number MFCD00005899