missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2129236

 View more versions of this product

Catalog No. AAB2129236



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-(Hydroxymethyl)phthalimide
140°C to 147°C
MFCD00005899
140946
MNSGOOCAMMSKGI-UHFFFAOYSA-N
2-(hydroxymethyl)isoindole-1,3-dione
8354
177.16
118-29-6
C9H7NO3
500 g
n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
C1=CC=C2C(=C1)C(=O)N(C2=O)CO
177.159
CHEBI:38816
97%

Chemical Identifiers

118-29-6
177.159
MNSGOOCAMMSKGI-UHFFFAOYSA-N
8354
2-(hydroxymethyl)isoindole-1,3-dione
C9H7NO3
MFCD00005899
n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
CHEBI:38816
C1=CC=C2C(=C1)C(=O)N(C2=O)CO
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 204-241-4

RTECSNumber : TI5270000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only