Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Hydroxymethyl)nonanamide, 98%

$40.47 - $621.86

Chemical Identifiers

Chemical Name or Material N-(Hydroxymethyl)nonanamide
CAS 130535-83-0
Molecular Formula CH3(CH2)7C(O)NHCH2OH
Formula Weight 187.29
MDL Number MFCD00156149
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1374309

  SDS
alfa aesar™
A1374309
10g Each for $40.47

AAA1374318

  SDS
alfa aesar™
A1374318
50g Each for $155.47

AAA1374330

  SDS
alfa aesar™
A1374330
250g Each for $621.86
Description & Specifications

Specifications

Formula Weight 187.29
Chemical Name or Material N-(Hydroxymethyl)nonanamide
Molecular Formula CH3(CH2)7C(O)NHCH2OH
CAS 130535-83-0
Quantity 10g
Assay 98%
MDL Number MFCD00156149
Boiling Point 83° to 87°C
Melting Point 83° to 87°C