Please login to your online account to display your discounted pricing

N-Hydroxybenzenesulfonamide 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC411940100

 View more versions of this product

Catalog No. AC411940100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Granular Crystalline Powder
Assay ≥96.0%
Formula Weight 173.19
Quantity 10g
Color White
TSCA TSCA
RTECS Number MX9350000
EINECS Number 209-973-8
Melting Point 120° to 125°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material N-Hydroxybenzenesulfonamide
Packaging Glass bottle
Name Note 97%
Molecular Formula C6H7NO3S
CAS 599-71-3
Infrared Spectrum Authentic

Specifications

Chemical Name or Material N-Hydroxybenzenesulfonamide
Name Note 97%
CAS 599-71-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Molecular Formula C6H7NO3S
Formula Weight 173.19