Please login to your online account to display your discounted pricing

N'-Hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material N'-Hydroxy-4-[(2-methyl-1, 3-thiazol-4-yl)methoxy]benzenecarboximidamide
CAS 306935-19-3
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H13N3O2S
Formula Weight 263.32
MDL Number MFCD00830779
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

JM00487DA

 
fisher scientific company
JM00487DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

JM00487CB

 
fisher scientific company
JM00487CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.32
Quantity 1g
Assay 95%
CAS Min % 95.0
Chemical Name or Material N'-Hydroxy-4-[(2-methyl-1, 3-thiazol-4-yl)methoxy]benzenecarboximidamide
Molecular Formula C12H13N3O2S
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
MDL Number MFCD00830779
CAS 306935-19-3
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.