Please login to your online account to display your discounted pricing

N'-Hydroxy-2-(trifluoromethoxy)benzenecarboximidamide, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
CAS 63968-84-3
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H7F3N2O2
Formula Weight 220.15
MDL Number MFCD07772878
Synonym N«-Hydroxy-2-(trifluoromethoxy)benzenecarboximidamide (IUPAC)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO01204DA

  SDS
fisher scientific company
MO01204DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Assay 95%
Formula Weight 220.15
CAS Min % 95.0
Packaging Amber Glass Bottle
Molecular Formula C8H7F3N2O2
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material N'-hydroxy-2-(trifluoromethoxy)benzenecarboximidamide
Assay Percent Range 95%
MDL Number MFCD07772878
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 63968-84-3
Synonym N«-Hydroxy-2-(trifluoromethoxy)benzenecarboximidamide (IUPAC)
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.