Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Hydroxy-1,8-naphthalimide sodium salt, 99%

$32.34 - $117.64

Chemical Identifiers

Chemical Name or Material N-Hydroxy-1,8-naphthalimide sodium salt
CAS 6207-89-2
Molecular Formula C12H6NNaO3
Formula Weight 235.18g/mol
MDL Number MFCD00006923
Beilstein 4282212
Synonym N,N-(1,8-Naphthalyl)hydroxylamine sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1602706

 
alfa aesar™
A16027-06
5g Each for $32.34

AAA1602714

 
alfa aesar™
A16027-14
25g Each for $117.64
Description & Specifications

Specifications

Melting Point >300°C
Quantity 5g
Chemical Name or Material N-Hydroxy-1,8-naphthalimide sodium salt
CAS 6207-89-2
Assay 99%
Molecular Formula C12H6NNaO3
Formula Weight 235.18g/mol
MDL Number MFCD00006923
Beilstein 4282212
Synonym N,N-(1,8-Naphthalyl)hydroxylamine sodium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.