Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Hydroxy-1,8-naphthalimide, 97%

$69.27 - $618.80

Chemical Identifiers

Chemical Name or Material N-Hydroxy-1,8-naphthalimide
CAS 7797-81-1
Molecular Formula C12H7NO3
Formula Weight 213.19
MDL Number MFCD00065062
Beilstein 178595
Synonym N,N-(1,8-Naphthalyl)hydroxylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2248506

  SDS
alfa aesar™
B2248506
5g Each for $69.27

AAB2248514

  SDS
alfa aesar™
B2248514
25g Each for $190.86

AAB2248522

  SDS
alfa aesar™
B2248522
100g Each for $618.80
Description & Specifications

Specifications

Formula Weight 213.19
Chemical Name or Material N-Hydroxy-1,8-naphthalimide
TSCA TSCA
Assay 97%
Molecular Formula C12H7NO3
MDL Number MFCD00065062
CAS 7797-81-1
Beilstein 178595
Synonym N,N-(1,8-Naphthalyl)hydroxylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 283° to 286°C
Quantity 5g

Applications
It is used in chemical research & chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.