Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexylsulfonylacetonitrile, 98%

$60.66 - $197.03

Chemical Identifiers

Chemical Name or Material n-Hexylsulfonylacetonitrile
CAS 203310-42-3
Molecular Formula C8H15NO2S
Formula Weight 189.28
MDL Number MFCD00041475
Synonym n-Hexanesulfonylacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1058106

  SDS
alfa aesar™
L1058106
5g Each for $197.03

AAL1058103

  SDS
alfa aesar™
L1058103
1g Each for $60.66
Description & Specifications

Specifications

Formula Weight 189.28
Chemical Name or Material n-Hexylsulfonylacetonitrile
Assay 98%
Molecular Formula C8H15NO2S
MDL Number MFCD00041475
CAS 203310-42-3
Synonym n-Hexanesulfonylacetonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Melting Point 41° to 43°C
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.