missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexylsulfonylacetonitrile, 98%

Manufacturer:  Alfa Aesar™ L1058106

 View more versions of this product

Catalog No. AAL1058106



Specifications

Specifications

n-Hexylsulfonylacetonitrile
5g
C8H15NO2S
2-hexylsulfonyl acetonitrile, n-hexylsulfonylacetonitrile, n-hexanesulphonylacetonitrile, hexane-1-sulfonylacetonitrile, 2-hexane-1-sulfonyl acetonitrile
CCCCCCS(=O)(=O)CC#N
189.273
189.28
41°C to 43°C
203310-42-3
MFCD00041475
CGKSNJYMJMDPQT-UHFFFAOYSA-N
2-hexylsulfonylacetonitrile
2817563
98%

Chemical Identifiers

203310-42-3
189.273
CGKSNJYMJMDPQT-UHFFFAOYSA-N
2817563
CCCCCCS(=O)(=O)CC#N
C8H15NO2S
MFCD00041475
2-hexylsulfonyl acetonitrile, n-hexylsulfonylacetonitrile, n-hexanesulphonylacetonitrile, hexane-1-sulfonylacetonitrile, 2-hexane-1-sulfonyl acetonitrile
2-hexylsulfonylacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents