missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Hexylphosphonic acid, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04412109

 View more versions of this product

Catalog No. AA4412109



Description

Description

n-Hexylphosphonic acid is used in the manufacturing of nanoparticles such as quantum dots, nano-metals and nano-ceramics. It is also used for coating of many materials, including nanoparticles by condensed hydrophobic monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Hexylphosphonic acid
108°C to 110°C
C6H15O3P
10 g
Insoluble in water.
CCCCCCP(=O)(O)O
166.157
166.16
4721-24-8
White
MFCD00013961
n-hexylphosphonic acid, phosphonic acid, p-hexyl, hexylphosphonicacid, p-hexyl-phosphonic acid, hexylphosphonic acid
GJWAEWLHSDGBGG-UHFFFAOYSA-N
hexylphosphonic acid
312552
Powder

Chemical Identifiers

4721-24-8
166.157
GJWAEWLHSDGBGG-UHFFFAOYSA-N
312552
CCCCCCP(=O)(O)O
C6H15O3P
MFCD00013961
n-hexylphosphonic acid, phosphonic acid, p-hexyl, hexylphosphonicacid, p-hexyl-phosphonic acid, hexylphosphonic acid
hexylphosphonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only