missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Hexyl Sulfide 95%, ACROS Organics™

Chemical Identifiers

CAS 6294-31-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12 H26 S
Linear Formula [CH3(CH2)5]2S
Formula Weight 202.4
MDL Number MFCD00009528
Chemical Name or Material n-Hexyl sulfide, 95%
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC120800250 SDS Thermo Fisher Scientific
120800250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6294-31-1
100.0
C12 H26 S
202.4
n-Hexyl sulfide, 95%
94.0
95%
[CH3(CH2)5]2S
MFCD00009528
≥94%

Specifications

Liquid
25g
0.8400g/mL
100.0
1.4560 to 1.4600 (20°C, 589nm)
Authentic
95%
C12 H26 S
n-Hexyl sulfide, 95%
202.4
Clear colorless
01, 409
94.0
Glass bottle
[CH3(CH2)5]2S
MFCD00009528
6294-31-1
≥94%

Safety and Handling

STENCH

EINECSNumber : 228-556-1

TSCA : TSCA