Learn More
n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
$56.66 - $152.48
Chemical Identifiers
CAS | 142-09-6 |
---|---|
Molecular Formula | C10H18O2 |
Molecular Weight (g/mol) | 170.25 |
MDL Number | MFCD00015283 |
InChI Key | LNCPIMCVTKXXOY-UHFFFAOYSA-N |
Synonym | hexyl methacrylate, n-hexyl methacrylate, hexyl 2-methyl-2-propenoate, methacrylic acid, hexyl ester, 2-propenoic acid, 2-methyl-, hexyl ester, unii-538b2q3hsv, ccris 4824, hexyl 2-methylacrylate, methacrylic acid hexyl ester, dsstox_cid_5406 |
PubChem CID | 8872 |
IUPAC Name | hexyl 2-methylprop-2-enoate |
SMILES | CCCCCCOC(=O)C(C)=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1156914
|
Thermo Scientific Chemicals
L1156914 |
25 g |
Each for $56.66
|
|
|||||
AAL1156922
|
Thermo Scientific Chemicals
L1156922 |
100 g |
Each for $152.48
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-09-6 | |
170.25 | |
LNCPIMCVTKXXOY-UHFFFAOYSA-N | |
8872 | |
CCCCCCOC(=O)C(C)=C |
C10H18O2 | |
MFCD00015283 | |
hexyl methacrylate, n-hexyl methacrylate, hexyl 2-methyl-2-propenoate, methacrylic acid, hexyl ester, 2-propenoic acid, 2-methyl-, hexyl ester, unii-538b2q3hsv, ccris 4824, hexyl 2-methylacrylate, methacrylic acid hexyl ester, dsstox_cid_5406 | |
hexyl 2-methylprop-2-enoate |
Specifications
142-09-6 | |
0.885 | |
80°C (176°F) | |
C10H18O2 | |
MFCD00015283 | |
1754703 | |
LNCPIMCVTKXXOY-UHFFFAOYSA-N | |
hexyl 2-methylprop-2-enoate | |
8872 | |
97% |
-57°C | |
204°C | |
Characteristic | |
1.431 | |
25 g | |
hexyl methacrylate, n-hexyl methacrylate, hexyl 2-methyl-2-propenoate, methacrylic acid, hexyl ester, 2-propenoic acid, 2-methyl-, hexyl ester, unii-538b2q3hsv, ccris 4824, hexyl 2-methylacrylate, methacrylic acid hexyl ester, dsstox_cid_5406 | |
CCCCCCOC(=O)C(C)=C | |
170.25 | |
170.25 | |
n-Hexyl methacrylate, stabilized with 100ppm 4-methoxyphenol |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 205-521-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only