Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexyl formate, 97%

Chemical Identifiers

Chemical Name or Material n-Hexyl formate
CAS 629-33-4
Molecular Formula CH3(CH2)5O2CH
Formula Weight 130.19
MDL Number MFCD00046146
UN Number UN3272
Beilstein 1701629
Synonym Formic acid n-hexyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0622618

  SDS
alfa aesar™
L0622618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0622630

  SDS
alfa aesar™
L0622630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.19
Chemical Name or Material n-Hexyl formate
Health Hazard 1 Warning
Molecular Formula CH3(CH2)5O2CH
CAS 629-33-4
TSCA TSCA
Beilstein 1701629
RTECS Number LQ8595000
Quantity 50g
MDL Number MFCD00046146
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Formic acid n-hexyl ester
UN Number UN3272
Boiling Point 155°C
Melting Point −63°C
Flash Point 37°C (98°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Assay 97%
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.