missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexyl formate, 97%

Manufacturer:  Alfa Aesar™ L0622618

 View more versions of this product

Catalog No. AAL0622618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

n-Hexyl formate
155°C
50g
C7H14O2
UN3272
n-hexyl formate, formic acid, hexyl ester, n-hexyl methanoate, hexyl methanoate, formic acid hexyl ester, hexyl formiate, unii-9gpu7pgv1g, fema no. 2570, 9gpu7pgv1g, hexylformate
CCCCCCOC=O
130.187
130.19
37°C (98°F)
-63°C
629-33-4
MFCD00046146
1701629
OUGPMNMLWKSBRI-UHFFFAOYSA-N
hexyl formate
61177
97%

Chemical Identifiers

629-33-4
130.187
OUGPMNMLWKSBRI-UHFFFAOYSA-N
61177
CCCCCCOC=O
C7H14O2
MFCD00046146
n-hexyl formate, formic acid, hexyl ester, n-hexyl methanoate, hexyl methanoate, formic acid hexyl ester, hexyl formiate, unii-9gpu7pgv1g, fema no. 2570, 9gpu7pgv1g, hexylformate
hexyl formate
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

RTECSNumber : LQ8595000

TSCA : TSCA

Documents