Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexyl acrylate, 95%, stab. with hydroquinone

Chemical Identifiers

Chemical Name or Material n-Hexyl acrylate
CAS 2499-95-8
Name Note stabilized with hydroquinone
Molecular Formula C9H16O2
Linear Formula H2C=CHCO2(CH2)5CH3
Formula Weight 156.22
MDL Number MFCD00048881
UN Number UN3082
Beilstein 1757327
Synonym Acrylic acid n-hexyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0990522

  SDS
alfa aesar™
L0990522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0990514

  SDS
alfa aesar™
L0990514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 156.22
MDL Number MFCD00048881
Assay 95%
Synonym Acrylic acid n-hexyl ester
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
RTECS Number AT1450000
Linear Formula H2C=CHCO2(CH2)5CH3
Quantity 100g
CAS 2499-95-8
Chemical Name or Material n-Hexyl acrylate
Molecular Formula C9H16O2
UN Number UN3082
TSCA TSCA
Flash Point 68°C (154°F)
Melting Point −45°C
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 1 Warning
Beilstein 1757327
Name Note stabilized with hydroquinone
Boiling Point 88° to 89°C (24mmHg)
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.