missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexyl acrylate, 95%, stab. with hydroquinone

Manufacturer:  Alfa Aesar™ L0990514

 View more versions of this product

Catalog No. AAL0990514



Specifications

Specifications

n-Hexyl acrylate
88°C to 89°C (24mmHg)
25g
2499-95-8
H2C=CHCO2(CH2)5CH3
UN3082
hexyl acrylate, n-hexyl acrylate, hexyl 2-propenoate, 2-propenoic acid, hexyl ester, n-hexylacrylate, ageflex n-ha, acrylic acid, hexyl ester, ccris 7038, acrylic acid-n-hexyl ester, hexylacrylate
CCCCCCOC(=O)C=C
156.225
156.22
68°C (154°F)
-45°C
stabilized with hydroquinone
C9H16O2
MFCD00048881
1757327
LNMQRPPRQDGUDR-UHFFFAOYSA-N
hexyl prop-2-enoate
17259
95%

Chemical Identifiers

2499-95-8
156.225
LNMQRPPRQDGUDR-UHFFFAOYSA-N
17259
CCCCCCOC(=O)C=C
C9H16O2
MFCD00048881
hexyl acrylate, n-hexyl acrylate, hexyl 2-propenoate, 2-propenoic acid, hexyl ester, n-hexylacrylate, ageflex n-ha, acrylic acid, hexyl ester, ccris 7038, acrylic acid-n-hexyl ester, hexylacrylate
hexyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

RTECSNumber : AT1450000

TSCA : TSCA

Documents