missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Hexyl acetate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L06342AU

 View more versions of this product

Catalog No. AAL06342AU



Description

Description

n-Hexyl acetate exhibits antimicrobial activity and can be used to improve the safety of minimally processed fruits. It is a fruity smelling fluid used as flavoring agent or in perfumes. It is a green leaf volatile from cabbage Brassica oleracea var. capitata L. It is used as plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Hexyl acetate exhibits antimicrobial activity and can be used to improve the safety of minimally processed fruits. It is a fruity smelling fluid used as flavoring agent or in perfumes. It is a green leaf volatile from cabbage Brassica oleracea var. capitata L. It is used as plasticizers.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store at 4°C. Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications

Specifications

n-Hexyl acetate
-80°C
167°C to 169°C
C8H16O2
CH3CO2(CH2)5CH3
1000 mL
1747138
Soluble in alcohol,ether. Insoluble in water.
CCCCCCOC(=O)C
144.214
CHEBI:87510
99%
142-92-7
0.873
43°C (109°F)
1.409
MFCD00009524
UN3271
n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
AOGQPLXWSUTHQB-UHFFFAOYSA-N
hexyl acetate
8908
144.21

Chemical Identifiers

142-92-7
144.214
AOGQPLXWSUTHQB-UHFFFAOYSA-N
8908
hexyl acetate
C8H16O2
MFCD00009524
n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
CHEBI:87510
CCCCCCOC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P374-P380-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 205-572-7

RTECSNumber : AI0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only