Please login to your online account to display your discounted pricing

n-Hexane, Spectrophotometric Grade, 95+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material n-Hexane
CAS 110-54-3
Purity Grade Spectrophotometric grade
Molecular Formula CH3(CH2)4CH3
Formula Weight 86.18
MDL Number MFCD00009520
UN Number UN1208
Beilstein 1730733
Merck Index 14,4694
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA32454K2

  SDS
alfa aesar
32454K2
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Spectrophotometric grade
Formula Weight 86.18
Quantity 1L
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1208
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H315-H336;
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Assay ≥95%
Flash Point −23°C (−9°F)
Merck Index 14,4694
Refractive Index 1.3750
Odor Weak, Characteristic
Boiling Point 69°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
Chemical Name or Material n-Hexane
CAS 110-54-3
RTECS Number MN9275000
Physical Form Liquid
Molecular Formula CH3(CH2)4CH3
MDL Number MFCD00009520
Beilstein 1730733
Density 0.659
Melting Point −95°C
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H361-H373-H315-H336;
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.