missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Hexane For spectroscopy Uvasol™, MilliporeSigma™
For spectroscopy Uvasol™
$135.24 - $482.26
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
M1043720500
|
Technidata
1043720500 |
500 mL | Glass Bottle |
N/A
|
|
||||
M1043722500
|
Technidata
1043722500 |
2.5 L | Glass Bottle |
N/A
|
|
||||
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-94.3°C | |
Colorless | |
69°C | |
−22°C (−7.6°F) | |
Benzine-like | |
Liquid | |
C6H14 | |
160 hPa (20°C) | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 |
110-54-3 | |
0.66g/cm3 at 20°C | |
500 mL | |
240° (464°F) | |
Glass Bottle | |
1.37 | |
MFCD02179311 | |
1208 | |
Soluble in water: 0.0095g/L at 20°C | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
Hexane |
Safety and Handling
H304: May be fatal if swallowed and enters airways.
H315: Causes skin irritation.
H225: Highly flammable liquid and vapour.
Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.