missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Hexane, EMD Millipore

Manufacturer:  Millipore Sigma HX02956

 View more versions of this product

Catalog No. MHX02956



Specifications

Specifications

n-Hexane
110-54-3
C6H14
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.178
CHEBI:29021
Glass Bottle
OmniPure™
95%
CH3(CH2)4CH3
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
1L

Chemical Identifiers

110-54-3
86.178
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC
C6H14
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

Documents