Please login to your online account to display your discounted pricing

n-Hexane-d14, for NMR, packaged in ampoules, 99% atom D, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  321390010

 View more versions of this product

Catalog No. AC321390010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 100.28
Quantity 1g
Color Clear colorless
Density 0.9600g/mL
Purity Grade For NMR
CAS 21666-38-6
Chemical Name or Material n-Hexane-d14
Name Note 99 atom% D, for NMR
Molecular Formula C6D14
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
H
Linear Formula D3C(CD2)4CD3
MDL Number MFCD00037562
Deuterium (D) 98.5% min.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material n-Hexane-d14
Name Note 99 atom% D, for NMR
CAS 21666-38-6
Purity Grade For NMR
Molecular Formula C6D14
Linear Formula D3C(CD2)4CD3
Formula Weight 100.28
MDL Number MFCD00037562
Accessories

GHS Signal Word: Danger

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
H

SDS & Safety Documents